Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102611.1 | NP_001096081.1 | 5 | 1122 | ||
GenBank mRNA | AK030380.1 | - | 5 | 959 | ||
AK089959.1 | - | 5 | 1058 | |||
AK154507.1 | - | 4 | 808 | |||
AK173306.1 | - | 3 | 544 | |||
BC049810.1 | - | 5 | 973 | |||
BC095952.1 | - | 5 | 1019 | 5037842 | ||
BC130220.1 | - | 2 | 459 | 40141368 |