Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177016.3 | NP_795990.2 | 12 | 1759 | ||
GenBank mRNA | AK033669.1 | - | 12 | 1719 | ||
AK050272.1 | - | 10 | 2808 | |||
AK162320.1 | - | 11 | 2202 |