Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177033.3 | NP_796007.1 | 4 | 1693 | ||
GenBank mRNA | AK033944.1 | - | 4 | 1487 | ||
AK044771.1 | - | 4 | 1228 | |||
AK141514.1 | - | 4 | 1694 | |||
BC120564.1 | - | 3 | 1155 | 40129487 | ||
BC137825.1 | - | 3 | 1156 | 8860844 | ||
DQ421811.1 | - | 3 | 1159 |