Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177034.3 | NP_796008.2 | 13 | 4025 | ||
GenBank mRNA | AK032261.1 | - | 12 | 2789 | ||
AK052243.1 | - | 1 | 483 | |||
AK147623.1 | - | 13 | 3977 | |||
BC141181.1 | - | 13 | 3867 | 9055793 |