Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177075.4 | NP_796049.1 | 2 | 924 | ||
GenBank mRNA | AK081240.1 | - | 2 | 925 |