Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252094.1 | NP_001239023.1 | 3 | 648 | ||
NM_001252095.1 | NP_001239024.1 | 3 | 629 | |||
NM_001252096.1 | NP_001239025.1 | 2 | 538 | |||
NM_177101.5 | NP_796075.2 | 2 | 519 | |||
GenBank mRNA | AK029471.1 | - | 2 | 479 | ||
AK077292.1 | - | 2 | 426 | |||
AK160463.1 | - | 3 | 627 | |||
BB614316.1 | - | 2 | 479 | |||
BC048711.1 | - | 2 | 411 | 6328428 | ||
BC065807.1 | - | 2 | 435 | 3373106 | ||
DN174886.1 | - | 2 | 412 |