Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnrc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144812.2 | NP_659061.2 | 5, 6 | 500 | ||
NM_177124.4 | NP_796098.3 | 2, 3 | 291 | |||
GenBank mRNA | AK048628.1 | - | 5, 6 | 500 | ||
AK051922.1 | - | 2, 3 | 260 | |||
BC068158.1 | - | 2, 3 | 186 | |||
CK639087.1 | - | 2, 3 | 291 | |||
CN699186.1 | - | 2, 3 | 212 |