Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fryl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028194.2 | NP_082470.2 | 8 | 1056 | ||
GenBank mRNA | AK033493.1 | - | 8 | 864 | ||
AK036035.1 | - | 6 | 774 | |||
AK164590.1 | - | 8 | 1057 |