Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177192.3 | NP_796166.2 | 3 | 632 | ||
GenBank mRNA | AK038858.1 | - | 3 | 534 | ||
AK050728.1 | - | 3 | 490 | |||
AK052296.1 | - | 3 | 487 | |||
AK135015.1 | - | 3 | 517 | |||
BC132199.1 | - | 3 | 337 | 40130476 | ||
BC132535.1 | - | 3 | 337 | 40130812 | ||
BC145128.1 | - | 3 | 337 |