Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177200.4 | NP_796174.2 | 4 | 299 | ||
GenBank mRNA | AK034997.1 | - | 4 | 300 | ||
AK137043.1 | - | 4 | 300 | |||
BB627230.1 | - | 4 | 300 | |||
BC058741.1 | - | 4 | 282 | 6309979 |