Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177115.2 | NP_796089.1 | 5 | 1078 | ||
GenBank mRNA | AK041088.1 | - | 5 | 1078 | ||
BC107232.1 | - | 5 | 862 | 40052073 | ||
BC107233.1 | - | 5 | 862 | 40052074 |