Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177140.3 | NP_796114.1 | 2 | 585 | ||
GenBank mRNA | AK032513.1 | - | 2 | 591 | ||
AK052027.1 | - | 2 | 587 | |||
AK133919.1 | - | 2 | 588 | |||
AK144149.1 | - | 588 | ||||
AK172131.1 | - | 2 | 210 | |||
BC096606.1 | - | 2 | 573 | 5366962 |