Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002924.2 | NP_001002924.2 | 3, 4 | 630 | ||
NM_001284.3 | NP_001275.1 | 2, 3 | 594 | |||
NM_001318091.1 | NP_001305020.1 | 2, 3 | 594 | |||
NM_001318093.1 | NP_001305022.1 | 2, 3 | 594 | |||
NM_001318094.1 | NP_001305023.1 | 2, 3 | 594 | |||
XM_011543136.2 | XP_011541438.2 | 2, 3 | 594 | |||
XM_017009024.1 | XP_016864513.1 | 2, 3 | 594 | |||
XM_017009025.1 | XP_016864514.1 | 2, 3 | 594 | |||
GenBank mRNA | AK311843.1 | - | 2, 3 | 162 | ||
BC000804.1 | - | 2, 3 | 246 | 3452302 | ||
BC012614.1 | - | 3, 4 | 243 | |||
BI600357.1 | - | 2, 3 | 282 | |||
BT009833.1 | - | 2, 3 | 162 | |||
BX394598.2 | - | 2, 3 | 173 | |||
CN411347.1 | - | 2, 3 | 275 | |||
CR450341.1 | - | 2, 3 | 162 | |||
D63643.1 | - | 2, 3 | 204 | |||
U91932.1 | - | 2, 3 | 247 | |||
X99458.1 | - | 2, 3 | 192 |