Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290138.1 | NP_001277067.1 | 5, 6 | 1086 | ||
NM_001290141.1 | NP_001277070.1 | 5, 6 | 561 | |||
NM_177203.3 | NP_796177.2 | 5, 6 | 465 | |||
GenBank mRNA | AK042903.1 | - | 5, 6 | 1088 | ||
BC150827.1 | - | 4, 5 | 283 | |||
BC171976.1 | - | 4, 5 | 283 | |||
BC171978.1 | - | 5, 6 | 317 | |||
BC171981.1 | - | 4, 5 | 283 | |||
BU962217.1 | - | 467 |