Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrig3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177152.5 | NP_796126.4 | 8 | 1239 | ||
GenBank mRNA | AK018522.1 | - | 2 | 52 | ||
AK035166.1 | - | 8 | 1241 | |||
AK137007.1 | - | 8 | 1226 | |||
AY505341.1 | - | 8 | 1182 | |||
BC065142.1 | - | 8 | 1205 | 30610561 |