Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Islr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161535.1 | NP_001155007.1 | 2 | 3765 | ||
NM_001161536.1 | NP_001155008.1 | 4 | 3946 | |||
NM_001161537.1 | NP_001155009.1 | 3 | 3937 | |||
NM_001161538.1 | NP_001155010.1 | 4 | 3714 | |||
NM_001161539.1 | NP_001155011.1 | 4 | 3676 | |||
NM_001161540.1 | NP_001155012.1 | 3 | 3632 | |||
NM_001161541.1 | NP_001155013.1 | 3 | 3523 | |||
NM_177193.5 | NP_796167.2 | 3 | 3756 | |||
GenBank mRNA | AK043849.1 | - | 1 | 1591 | ||
AK047348.1 | - | 1 | 3244 | |||
AK129367.1 | - | 3 | 3443 | |||
BC059068.1 | - | 3 | 3609 | 6825665 | ||
BC096531.1 | - | 2 | 3765 | 6826287 |