Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034997.1 | - | 11 | 1398 | ||
AK036830.1 | - | 1 | 1808 | |||
AK137043.1 | - | 11 | 1398 |