Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ocrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177215.3 | NP_796189.2 | 2, 3 | 309 | ||
GenBank mRNA | AK035315.1 | - | 2, 3 | 312 | ||
AK041963.1 | - | 2, 3 | 311 | |||
AK047906.1 | - | 2, 3 | 328 | |||
AK138269.1 | - | 2, 3 | 311 | |||
BC068146.1 | - | 2, 3 | 309 | 30533468 |