Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177256.5 | NP_796230.1 | 1 | 705 | ||
GenBank mRNA | AK029613.1 | - | 1 | 664 | ||
AK077012.1 | - | 1 | 701 | |||
AK161395.1 | - | 1 | 703 | |||
BC087895.1 | - | 1 | 692 | 6771677 |