Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177260.2 | NP_796234.1 | 2 | 243 | ||
GenBank mRNA | AK037111.1 | - | 2 | 155 | ||
AK081441.1 | - | 2 | 243 | |||
AK154236.1 | - | 2 | 158 | |||
AK170385.1 | - | 2 | 111 | |||
AK170626.1 | - | 2 | 58 | |||
AK170665.1 | - | 2 | 91 | |||
AK171510.1 | - | 2 | 226 | |||
BC096432.1 | - | 2 | 126 | 5390573 | ||
BC113174.1 | - | 2 | 183 | 40060792 | ||
BC113175.1 | - | 2 | 183 | 40060793 |