Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Negr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039094.3 | NP_001034183.1 | 1, 2 | 567 | ||
NM_177274.4 | NP_796248.1 | 1, 2 | 567 | |||
GenBank mRNA | AJ487032.1 | - | 1, 2 | 187 | ||
AK030503.1 | - | 1, 2 | 393 | |||
AK035218.1 | - | 1, 2 | 403 | |||
AK142298.1 | - | 1, 2 | 261 | |||
AK147253.1 | - | 1, 2 | 568 | |||
BC146005.1 | - | 1, 2 | 352 | 40131308 | ||
BC146007.1 | - | 1, 2 | 351 | 40131310 |