Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wscd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177292.3 | NP_796266.2 | 1283 | |||
GenBank mRNA | AK049685.1 | - | 979 | |||
AK173026.1 | - | 1 | 1227 | |||
BC096526.1 | - | 1283 | ||||
BC147280.1 | - | 1 | 311 | 9056352 | ||
BC147281.2 | - | 1 | 311 | 9056324 |