Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700006J14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005678.1 | - | 3 | 611 | ||
AK011105.1 | - | 4 | 651 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177313.1 | - | 611 | Chr. - - [-] on Build |