Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amigo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177275.4 | NP_796249.1 | 1 | 628 | ||
GenBank mRNA | AB167510.1 | - | 1 | 576 | ||
AK028619.1 | - | 1 | 576 | |||
AK088051.1 | - | 1 | 629 | |||
AY237004.1 | - | 1 | 407 |