Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293312.1 | NP_001280241.1 | 5, 6 | 662 | ||
NM_001349.3 | NP_001340.2 | 6, 7 | 720 | |||
XM_017003489.1 | XP_016858978.1 | 6, 7 | 691 | |||
GenBank mRNA | AK129521.1 | - | 6, 7 | 662 | ||
AK222476.1 | - | 6, 7 | 550 | |||
AK290607.1 | - | 6, 7 | 577 | |||
AY762100.1 | - | 6, 7 | 539 | |||
BC000629.2 | - | 6, 7 | 553 | 3344322 | ||
BC107749.1 | - | 6, 7 | 533 | 6709590 | ||
BI753718.1 | - | 6, 7 | 692 | |||
BT006710.1 | - | 6, 7 | 504 | |||
CR749809.1 | - | 5, 6 | 470 | |||
J05032.1 | - | 6, 7 | 594 |