Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177304.3 | NP_796278.1 | 2 | 446 | ||
GenBank mRNA | AK038460.1 | - | 2 | 418 | ||
AK046797.1 | - | 2 | 447 | |||
AK046881.1 | - | 2 | 451 | |||
AK079352.1 | - | 2 | 428 | |||
AK079642.1 | - | 2 | 446 | |||
BC096376.1 | - | 2 | 417 | 4237600 |