Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177213.3 | NP_796187.2 | 6, 7 | 1019 | ||
GenBank mRNA | AK029712.1 | - | 6, 7 | 1019 | ||
AY243471.1 | - | 6, 7 | 1019 | |||
BC141350.1 | - | 6, 7 | 984 | 9055962 | ||
BC141351.1 | - | 6, 7 | 984 | 9055963 |