Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ocrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177215.3 | NP_796189.2 | 12 | 1366 | ||
GenBank mRNA | AK035315.1 | - | 12 | 1369 | ||
AK041963.1 | - | 12 | 1368 | |||
AK044320.1 | - | 11 | 1288 | |||
AK047906.1 | - | 12 | 1385 | |||
BC068146.1 | - | 12 | 1366 | 30533468 |