Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177271.3 | NP_796245.2 | 2 | 1659 | ||
GenBank mRNA | AK045645.1 | - | 2 | 1211 | ||
AK087498.1 | - | 2 | 1661 | |||
AK132444.1 | - | 2 | 1633 | |||
BC145672.1 | - | 2 | 1433 | 40130975 | ||
BC145674.1 | - | 2 | 1433 | 40130977 |