Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177304.3 | NP_796278.1 | 2 | 449 | ||
GenBank mRNA | AK038460.1 | - | 2 | 421 | ||
AK046797.1 | - | 2 | 450 | |||
AK046881.1 | - | 2 | 454 | |||
AK079352.1 | - | 2 | 431 | |||
AK079642.1 | - | 2 | 449 | |||
BC096376.1 | - | 2 | 420 | 4237600 |