Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177338.5 | NP_796312.2 | 10 | 1554 | ||
GenBank mRNA | AK089782.1 | - | 10 | 1555 |