Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290491.1 | NP_001277420.1 | 7 | 1308 | ||
NM_001290492.1 | NP_001277421.1 | 7 | 1316 | |||
NM_001290493.1 | NP_001277422.1 | 6 | 921 | |||
NM_177370.4 | NP_796344.1 | 7 | 1304 | |||
GenBank mRNA | AK053223.1 | - | 7 | 1143 | ||
AK077457.1 | - | 7 | 1140 | |||
AK156451.1 | - | 7 | 1158 | |||
BC062155.1 | - | 5 | 1023 | |||
BC096693.1 | - | 6 | 930 | 5255059 | ||
BC132459.1 | - | 6 | 969 | 40130736 | ||
BC138479.1 | - | 6 | 969 | 8861499 | ||
BC145287.1 | - | 6 | 981 | 9053841 | ||
BC145289.1 | - | 6 | 973 | 9053843 | ||
BC145290.1 | - | 6 | 973 |