Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290563.1 | NP_001277492.1 | 5 | 644 | ||
NM_001290564.1 | NP_001277493.1 | 5 | 699 | |||
NM_001290566.1 | NP_001277495.1 | 4 | 397 | |||
NM_001290567.1 | NP_001277496.1 | 5 | 445 | |||
NM_177384.3 | NP_796358.2 | 5 | 644 | |||
GenBank mRNA | AK028040.1 | - | 5 | 699 | ||
AK089384.1 | - | 4 | 677 | |||
AK133273.1 | - | 5 | 422 | |||
AK160435.1 | - | 5 | 395 | |||
AK164721.1 | - | 5 | 598 | |||
BC140991.1 | - | 4 | 388 | 9055603 | ||
BC145064.1 | - | 5 | 436 | 9053612 |