Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177331.4 | NP_796305.3 | 2, 3 | 396 | ||
GenBank mRNA | AK030995.1 | - | 2, 3 | 406 | ||
AK031077.1 | - | 2, 3 | 375 | |||
BC090653.1 | - | 2, 3 | 396 | 30659763 | ||
BC138157.1 | - | 2, 3 | 283 | 8861176 | ||
BC138158.1 | - | 2, 3 | 283 | 8861177 |