Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197293.2 | NP_001184222.1 | 8, 9 | 1266 | ||
NM_001244604.1 | NP_001231533.1 | 8, 9 | 908 | |||
NM_001386.5 | NP_001377.1 | 8, 9 | 1249 | |||
GenBank mRNA | AB209195.1 | - | 8, 9 | 987 | ||
AK223557.1 | - | 8, 9 | 1112 | |||
AK291287.1 | - | 8, 9 | 1030 | |||
AK299077.1 | - | 8, 9 | 901 | |||
BC056408.1 | - | 8, 9 | 1103 | 6044141 | ||
BC067109.1 | - | 8, 9 | 1133 | 3870039 | ||
BX647115.1 | - | 8, 9 | 1177 | |||
D78013.1 | - | 8, 9 | 1092 | |||
U17279.1 | - | 8, 9 | 889 |