Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177352.4 | NP_796326.1 | 4 | 370 | ||
GenBank mRNA | AK049275.1 | - | 4 | 371 | ||
AK076293.1 | - | 4 | 398 | |||
AK160477.1 | - | 4 | 425 | |||
AK162119.1 | - | 4 | 371 | |||
AK163031.1 | - | 4 | 366 | |||
AK167217.1 | - | 4 | 390 | |||
BC116302.1 | - | 4 | 367 | 40095102 | ||
BC116303.1 | - | 3 | 273 | 40095106 |