Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177382.4 | NP_796356.2 | 4 | 1066 | ||
GenBank mRNA | AK050453.1 | - | 4 | 416 | ||
AY323818.1 | - | 4 | 1066 | |||
BC108961.1 | - | 4 | 1066 | 40046632 | ||
BC108962.1 | - | 4 | 490 |