Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ust | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177387.3 | NP_796361.2 | 6 | 1168 | ||
GenBank mRNA | AK029782.1 | - | 5 | 559 | ||
AK086181.1 | - | 6 | 1115 | |||
AK147850.1 | - | 6 | 1169 | |||
AK162950.1 | - | 6 | 741 | |||
BC138155.1 | - | 6 | 908 | 8861174 |