Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164785.1 | NP_001158257.1 | 4 | 959 | ||
NM_001164786.1 | NP_001158258.1 | 4 | 959 | |||
NM_177431.4 | NP_803180.3 | 4 | 959 | |||
GenBank mRNA | AJ512753.3 | - | 4 | 728 | ||
AK090358.1 | - | 4 | 924 | |||
AK145288.1 | - | 4 | 960 | |||
AY189815.1 | - | 4 | 867 | |||
AY189816.1 | - | 4 | 867 | |||
BC121792.1 | - | 4 | 798 | 40099393 |