Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177546.2 | NP_808214.1 | 4 | 612 | ||
NM_211138.1 | NP_997593.1 | 4 | 623 | |||
GenBank mRNA | AK032027.1 | - | 4 | 325 | ||
AK053577.1 | - | 4 | 597 | |||
AK149304.1 | - | 4 | 613 | |||
AY189949.1 | - | 4 | 460 | |||
AY189950.1 | - | 4 | 612 | |||
BB642464.1 | - | 4 | 613 | |||
BC048917.1 | - | 4 | 494 | 6309383 |