Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177337.3 | NP_796311.2 | 2 | 951 | ||
GenBank mRNA | AK008804.3 | - | 2 | 860 | ||
AK050053.1 | - | 2 | 862 | |||
AK155765.1 | - | 1 | 1573 | |||
BC064093.1 | - | 2 | 951 | 864589 |