Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177366.3 | NP_796340.2 | 4 | 1586 | ||
GenBank mRNA | AK089429.1 | - | 4 | 1727 | ||
AK089544.1 | - | 4 | 1539 | |||
AK154291.1 | - | 4 | 1223 | |||
AK170941.1 | - | 4 | 1181 | |||
AK171081.1 | - | 4 | 1181 | |||
BC117996.1 | - | 4 | 1540 | 40095824 |
Set Membership: |
G-protein coupled receptor |