Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ust | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177387.3 | NP_796361.2 | 8 | 1734 | ||
GenBank mRNA | AK029782.1 | - | 7 | 1125 | ||
AK086181.1 | - | 8 | 1682 | |||
AK147850.1 | - | 8 | 1739 | |||
AK162950.1 | - | 8 | 1307 |