Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177561.3 | NP_808229.1 | 9 | 2537 | ||
GenBank mRNA | AK039074.1 | - | 1 | 1189 | ||
AK145298.1 | - | 9 | 2587 | |||
BC030888.1 | - | 7 | 2304 | |||
BC039916.1 | - | 9 | 2542 | 4950034 |