Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024937.3 | NP_001020108.1 | 4 | 456 | ||
NM_001321236.1 | NP_001308165.1 | 5 | 572 | |||
NM_015716.4 | NP_056531.1 | 4 | 456 | |||
NM_153827.4 | NP_722549.2 | 4 | 456 | |||
NM_170663.4 | NP_733763.1 | 4 | 456 | |||
XM_006721531.2 | XP_006721594.1 | 4 | 456 | |||
XM_006721532.2 | XP_006721595.1 | 4 | 456 | |||
XM_006721536.2 | XP_006721599.1 | 4 | 456 | |||
XM_011523906.1 | XP_011522208.1 | 4 | 456 | |||
XM_017024704.1 | XP_016880193.1 | 4 | 456 | |||
XM_017024705.1 | XP_016880194.1 | 4 | 456 | |||
XM_017024706.1 | XP_016880195.1 | 4 | 456 | |||
XM_017024707.1 | XP_016880196.1 | 4 | 257 | |||
XM_017024708.1 | XP_016880197.1 | 2 | 107 | |||
GenBank mRNA | AB035698.1 | - | 4 | 212 | ||
AB041926.1 | - | 4 | 396 | |||
AB209450.1 | - | 4 | 397 | |||
AY775058.1 | - | 4 | 212 | |||
BC034673.1 | - | 4 | 408 | 4384442 | ||
CA392419.1 | - | 4 | 429 | |||
DA653105.1 | - | 4 | 456 |