Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127197.1 | NP_001120669.1 | 3 | 540 | ||
NM_001421.3 | NP_001412.1 | 3 | 622 | |||
XM_005262389.3 | XP_005262446.1 | 3 | 958 | |||
XM_011531307.2 | XP_011529609.1 | 2 | 368 | |||
XM_011531308.2 | XP_011529610.1 | 1 | 83 | |||
GenBank mRNA | AF000670.1 | - | 2 | 339 | ||
AK314972.1 | - | 3 | 539 | |||
BC017194.2 | - | 3 | 505 | 3138355 | ||
CN314421.1 | - | 3 | 537 | |||
U32645.1 | - | 3 | 619 |