Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177589.3 | NP_808257.2 | 32, 33 | 3393 | ||
GenBank mRNA | AK043477.1 | - | 2, 3 | 968 | ||
AK132731.1 | - | 32, 33 | 3394 | |||
BC065094.1 | - | 33, 34 | 3691 |