Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177600.4 | NP_808268.2 | 13 | 1169 | ||
GenBank mRNA | AK029862.1 | - | 5 | 376 | ||
BC095976.1 | - | 12 | 915 | |||
BC150667.1 | - | 4 | 298 | 9087578 |