Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164376.1 | NP_001157848.1 | 2, 3 | 266 | ||
NM_177612.3 | NP_808280.2 | 2, 3 | 194 | |||
GenBank mRNA | AF344871.1 | - | 2, 3 | 266 | ||
AK030166.1 | - | 2, 3 | 195 | |||
AK142745.1 | - | 2, 3 | 267 | |||
BC112409.1 | - | 2, 3 | 168 | 40060582 | ||
DQ380429.1 | - | 2, 3 | 194 |