Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161365.1 | NP_001154837.1 | 2 | 173 | ||
NM_177620.4 | NP_808288.2 | 2 | 284 | |||
GenBank mRNA | AK032205.1 | - | 2 | 285 | ||
AK133988.1 | - | 2 | 192 | |||
AK154506.1 | - | 2 | 311 | |||
AK154522.1 | - | 2 | 88 | |||
AK154963.1 | - | 2 | 174 | |||
AK166196.1 | - | 2 | 130 | |||
AK167187.1 | - | 2 | 191 | |||
AK171301.1 | - | 2 | 97 | |||
AK171311.1 | - | 2 | 88 | |||
BC137907.1 | - | 2 | 212 | 8860926 |